LIF372156104 Life Chemicals Inc.

3-(2-fluorobenzyl)-3,6-dihydro-7H-[1,2,3]triazolo[4,5-d]pyrimidin-7-one

  • MDL number MFCD05253318
  • Cas Number : 119222-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart