LIF372156135 Life Chemicals Inc.

5-(3-bromophenyl)imidazolidine-2,4-dione

  • MDL number MFCD16631745
  • Cas Number : 79422-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart