LIF372156148 Life Chemicals Inc.

(5-bromo-2-methoxyphenyl)[(tert-butoxycarbonyl)amino]acetic acid

  • MDL number MFCD16653115
  • Cas Number : 1214038-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart