LIF372156223 Life Chemicals Inc.

4-(bromomethyl)-1-methoxy-2-nitrobenzene

  • MDL number MFCD03840698
  • Cas Number : 61010-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart