LIF372156232 Life Chemicals Inc.

1-[2-(dimethylamino)ethyl]-5-oxopyrrolidine-3-carboxamide

  • MDL number MFCD16631780
  • Cas Number : 1219827-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart