LIF372156233 Life Chemicals Inc.

4-isopropoxy-3,5-dimethoxybenzonitrile

  • MDL number MFCD16631781
  • Cas Number : 1219828-05-3
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart