LIF372156234 Life Chemicals Inc.

4-hydroxy-3,5-dimethoxybenzonitrile

  • MDL number MFCD09835372
  • Cas Number : 72684-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart