LIF372156242 Life Chemicals Inc.

4-(4-hydroxyphenyl)pyrrolidin-2-one

  • MDL number MFCD15526989
  • Cas Number : 55656-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart