LIF372156319 Life Chemicals Inc.

4-(aminomethyl)-1-cyclopropylpyrrolidin-2-one

  • MDL number MFCD09055283
  • Cas Number : 936940-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart