LIF372156336 Life Chemicals Inc.

1-methyl-5-oxopyrrolidine-3-carbonitrile

  • MDL number MFCD18819188
  • Cas Number : 172261-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart