LIF372156371 Life Chemicals Inc.

3-acetyl-6-chloro-4-(4-ethoxyphenyl)quinolin-2(1H)-one

  • MDL number MFCD21091974
  • Cas Number : 1283109-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart