LIF372156374 Life Chemicals Inc.

3-acetyl-6-chloro-4-(3,4-difluorophenyl)quinolin-2(1H)-one

  • MDL number MFCD21091977
  • Cas Number : 1283108-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart