LIF372156379 Life Chemicals Inc.

1-cyclopropyl-3-phenylpiperazine-2,5-dione

  • MDL number MFCD20877889
  • Cas Number : 1283107-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart