LIF372156391 Life Chemicals Inc.

7,7-dimethyl-6,8-dioxa-2-thiaspiro[3.5]nonane

  • MDL number MFCD21091986
  • Cas Number : 156720-48-8
  • Molecular Weight : 174.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart