LIF372156449 Life Chemicals Inc.

3-(4-fluorophenyl)azepane

  • MDL number MFCD21092011
  • Cas Number : 1333960-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart