LIF372156474 Life Chemicals Inc.

N-(1-phenylethyl)ethane-1,2-diamine

  • MDL number MFCD11593972
  • Cas Number : 99516-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart