LIF372156478 Life Chemicals Inc.

(3E)-3-[4-(trifluoromethyl)benzylidene]pyrrolidin-2-one

  • MDL number MFCD22587844
  • Cas Number : 1350994-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart