LIF372156489 Life Chemicals Inc.

4-(6-methylpyridin-2-yl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine

  • MDL number MFCD22587850
  • Cas Number : 1350988-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart