LIF372156503 Life Chemicals Inc.

3-(propan-2-yl)pyrrolidine

  • MDL number MFCD16681269
  • Cas Number : 34971-73-8
  • Molecular Weight : 113.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart