LIF372156504 Life Chemicals Inc.

3-tert-butylpyrrolidine

  • MDL number MFCD13192713
  • Cas Number : 381665-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart