LIF372156509 Life Chemicals Inc.

4-chloro-5-methoxy-2-methylpyrimidine

  • MDL number MFCD00223701
  • Cas Number : 698-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart