LIF372156521 Life Chemicals Inc.

1-(2-thienylcarbonyl)azetidine-3-carboxylic acid

  • MDL number MFCD20282281
  • Cas Number : 1339416-02-2
  • Molecular Weight : 211.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart