LIF372156524 Life Chemicals Inc.

1-[(2-fluorophenoxy)acetyl]azetidine-3-carboxylic acid

  • MDL number MFCD20550362
  • Cas Number : 1350989-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart