LIF372156538 Life Chemicals Inc.

5-oxo-1-(tetrahydro-2H-pyran-4-yl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD12088286
  • Cas Number : 1155632-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart