LIF372156539 Life Chemicals Inc.

4-amino-1-(4-chlorophenyl)pyrrolidin-2-one

  • MDL number MFCD12190545
  • Cas Number : 60655-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart