LIF372156540 Life Chemicals Inc.

4-chloro-5-fluoro-1H-indole-2,3-dione

  • MDL number MFCD03618550
  • Cas Number : 84378-94-9
  • Molecular Weight : 199.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-5-fluoro-1H-indole-2,3-dione
Inchi Code 1S/C8H3ClFNO2/c9-6-3(10)1-2-4-5(6)7(12)8(13)11-4/h1-2H,(H,11,12,13)
Inchi Key SWUGYKGLXFMLEN-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart