LIF372156550 Life Chemicals Inc.

1-(4-Amino-phenyl)-piperidin-2-one

  • MDL number MFCD09743291
  • Cas Number : 438056-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart