LIF372156581 Life Chemicals Inc.

5-methyl-2-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidin-7-amine

  • MDL number MFCD02725409
  • Cas Number : 380340-45-4
  • Molecular Weight : 217.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart