LIF372156585 Life Chemicals Inc.

(3-amino-4-chlorophenyl)(4-ethoxyphenyl)methanone

  • MDL number MFCD11604566
  • Cas Number : 71969-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart