LIF372156590 Life Chemicals Inc.

5-methyl-2-phenyl-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridin-3-amine

  • MDL number MFCD11574116
  • Cas Number : 1082387-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart