LIF372156594 Life Chemicals Inc.

1-(3,4-Dichloro-benzyl)-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid

  • MDL number MFCD00139964
  • Cas Number : 64488-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart