LIF372156595 Life Chemicals Inc.

2-(4-Benzylpiperidin-1-yl)quinolin-8-ol

  • MDL number MFCD09240616
  • Cas Number : 941868-41-3
  • Molecular Weight : 318.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart