LIF372156602 Life Chemicals Inc.

Ethyl 2-amino-4,5,6,7-tetrahydro-1,3-benzothiazole-4-carboxylate

  • MDL number MFCD08689777
  • Cas Number : 76263-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart