LIF372156603 Life Chemicals Inc.

2-(Benzoylamino)-5,6-dihydro-4H-cyclopenta[d][1,3]thiazole-4-carboxylic acid

  • MDL number MFCD09864119
  • Cas Number : 1019117-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart