LIF372156604 Life Chemicals Inc.

2-(4-fluorobenzamido)-5,6-dihydro-4H-cyclopenta[d]thiazole-4-carboxylic acid

  • MDL number MFCD09864120
  • Cas Number : 1019115-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart