LIF372156623 Life Chemicals Inc.

4-{[(3,5-dimethoxybenzyl)thio]methyl}-1,3-thiazol-2-amine

  • MDL number MFCD09864142
  • Cas Number : 1019111-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart