LIF372156627 Life Chemicals Inc.

2-{[(4-methoxyphenyl)sulfonyl]amino}-4-methyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD09864145
  • Cas Number : 951921-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart