LIF372156644 Life Chemicals Inc.

2-methoxy-4-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]phenol

  • MDL number MFCD09864158
  • Cas Number : 118828-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart