LIF372156654 Life Chemicals Inc.

4-Benzyloxybenzylamine

  • MDL number MFCD03410962
  • Cas Number : 22171-15-9
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart