LIF372156660 Life Chemicals Inc.

(2E)-4-(1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)but-2-enoic acid

  • MDL number MFCD11518886
  • Cas Number : 10295-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart