LIF372156665 Life Chemicals Inc.

2-({[(4-fluorophenyl)amino]carbonyl}amino)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD11986871
  • Cas Number : 1105191-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart