LIF372156669 Life Chemicals Inc.

4-Thiocarbamoylmethyl-piperazine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD11007745
  • Cas Number : 294622-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart