LIF372156683 Life Chemicals Inc.

2-(2-Furoylamino)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD09992841
  • Cas Number : 1003395-70-7
  • Molecular Weight : 238.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart