LIF372156689 Life Chemicals Inc.

(2-{[4-(trifluoromethoxy)phenyl]amino}-1,3-thiazol-4-yl)acetic acid

  • MDL number MFCD11986878
  • Cas Number : 918793-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart