LIF372156695 Life Chemicals Inc.

2-Chloromethyl-5-methoxy-pyran-4-one

  • MDL number MFCD04122657
  • Cas Number : 40838-34-4
  • Molecular Weight : 174.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart