LIF372156697 Life Chemicals Inc.

5-(benzyloxy)-4-oxo-4H-pyran-2-carboxylic acid

  • MDL number MFCD08531038
  • Cas Number : 1219-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart