LIF372156709 Life Chemicals Inc.

(4-Methanesulfonyl-phenyl)-thiourea

  • MDL number MFCD10693660
  • Cas Number : 84783-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart