LIF372156713 Life Chemicals Inc.

1-(2-Amino-6,7-dihydro-4H-thiazolo[5,4-c]pyridin-5-yl)-ethanone

  • MDL number MFCD10010939
  • Cas Number : 124458-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart