LIF372156735 Life Chemicals Inc.

[1-(4-fluorobenzyl)-1H-indol-3-yl]acetic acid

  • MDL number MFCD00794686
  • Cas Number : 176204-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart