LIF372156736 Life Chemicals Inc.

[1-(4-chlorobenzyl)-1H-indol-3-yl]acetic acid

  • MDL number MFCD09028380
  • Cas Number : 219544-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart